missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclopropanecarbonitrile, 97%, Thermo Scientific Chemicals

290.94 DKK - 839.81 DKK

Chemical Identifiers

CAS 935-44-4
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00001270
InChI Key ZHFURHRJUWYDKG-UHFFFAOYSA-N
Synonym 1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
PubChem CID 70288
IUPAC Name 1-phenylcyclopropane-1-carbonitrile
SMILES N#CC1(CC1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10174280
SDS View Documents
Thermo Scientific Acros
170590100
10 g Glass bottle
290.94 DKK
10g
Please to purchase this item. Need a web account? Register with us today!
10184280
SDS View Documents
Thermo Scientific Acros
170590500
50 g Glass bottle
839.81 DKK
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

935-44-4
143.19
ZHFURHRJUWYDKG-UHFFFAOYSA-N
70288
N#CC1(CC1)C1=CC=CC=C1
C10H9N
MFCD00001270
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
1-phenylcyclopropane-1-carbonitrile

Specifications

935-44-4
100.0
1.0000g/mL
>110°C
96% min. (GC)
C6H5C3H4CN
MFCD00001270
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
N#CC1(CC1)C1=CC=CC=C1
143.19
143.19
Liquid
96.0
Colorless to Yellow
133.0°C to 137.0°C (30.0 mmHg)
Authentic
C10H9N
1.5370 to 1.5400
1
ZHFURHRJUWYDKG-UHFFFAOYSA-N
1-phenylcyclopropane-1-carbonitrile
70288
97%
1-Phenyl-1-cyclopropanecarbonitrile, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 213-304-5

missing translation for 'tsca' : TSCA

Product Identifier
  • 1-Phenyl-1-cyclopropanecarbonitrile

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.