Learn More
Hamilton™ 10 μL 1700 Series Gas Tight Syringe
Product Code. p-200000261
Click to view available options
Needle Gauge:
22s G
23s G
26S (No Needle Included)
26s G
Custom
No Needle Included
Needle Point Style:
Blunt
Cone
Curved, Beveled (No Needle Included)
Custom
No Needle Included
Sharp, Bevel, Curved, Non-coring
Connection Type:
Cemented
Removable
Unit Size:
Each
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
  • This series of syringes are ideal for dispensing volumes up to 10 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Autoclavable Not Autoclavable
Material PTFE
Needle Gauge 23s G
Diameter (English) Inner 0.018 in.
Diameter (English) Outer 0.260 in.
Diameter (Metric) Inner 0.460 mm
Diameter (Metric) Outer 6.60 mm
Length (English) Needle 1.69 in.
Product Type Gas Tight Syringe
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 1701 SN Syringe
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 10 μL
Length (Metric) Needle 43 mm
Needle Point Style Custom
Connection Type Cemented