Learn More
SPEX Certiprep 1ML Benzidines Mix, 2,000 µg/mL
Brand:  SPEX Certiprep 605-X-MC
1ML Benzidines Mix, 2,000 µg/mL
Product Code. 16359817
  • 1425.00 DKK / 1mL
Explore more special offers
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Benzidines Mix
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : dichloromethane, benzidine, 3,3'-dichlorobenzidine