Learn More
2-Amino-6-chloro-3-nitropyridine, 98%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 192°C to 196°C |
Color | Yellow |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 474607 |
Solubility Information | Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform. |
Formula Weight | 173.56 |
Percent Purity | 98% |
Physical Form | Crystalline |
Chemical Name or Material | 2-Amino-6-chloro-3-nitropyridine |