Learn More
2-Mesitylethanol, 98%, Thermo Scientific™
Product Code. 11307965
Click to view available options
Quantity:
1 g
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6950-92-1
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00020609
InChI Key FQZPTDPHVFTOSY-UHFFFAOYSA-N
Synonym 2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol
PubChem CID 81389
IUPAC Name 2-(2,4,6-trimethylphenyl)ethanol
SMILES CC1=CC(C)=C(CCO)C(C)=C1
Specifications
Melting Point 80°C to 84°C
Quantity 25 g
Beilstein 1938400
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 2-Mesitylethanol