Learn More
Branson Ultrasonics™ 200- and 400-Watt Sonifiers: Wrenches
Wrenches for Branson 200- and 400-Watt Sonifiers
Brand:  Branson Ultrasonics™ 201-118-010
216.70 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11476218
  • 255.00 DKK / Each
Only null left
Estimated Shipment: 05-09-2025
Description
For use with Branson 200- and 400-Watt Sonifiers
Specifications
Specifications
Wrench
Titanium
Ideal for installing tip on tapped horns
0.5 in. tip