Learn More
3-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid, 97%, Thermo Scientific™
Product Code. 15400556
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 850568-50-2
Molecular Formula C10H13BO4
Molecular Weight (g/mol) 208.02
MDL Number MFCD04973736
InChI Key NSDRSKQWUQJUAE-UHFFFAOYSA-N
Synonym 3-2-methyl-1,3-dioxolan-2-yl phenylboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 2-3-boronobenzene-2-methyl-1,3-dioxolane, 2-3-boronophenyl-2-methyl-1,3-dioxolane, 3-2-methyl-1,3-dioxolan-2-yl benzeneboronic acid, 3-boronoacetophenone ethylene acetal, boronic acid,b-3-2-methyl-1,3-dioxolan-2-yl phenyl
PubChem CID 3674880
IUPAC Name [3-(2-methyl-1,3-dioxolan-2-yl)phenyl]boronic acid
SMILES CC1(OCCO1)C1=CC=CC(=C1)B(O)O
Specifications
Melting Point 100°C to 104°C
Quantity 1 g
Formula Weight 208.02
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid