Learn More
(3,3,3-Trifluoropropyl)methyldichlorosilane, 97%, Thermo Scientific Chemicals
Product Code. 11399305
Click to view available options
Quantity:
5 g
25 g
100 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 675-62-7
Molecular Formula C4H7Cl2F3Si
Molecular Weight (g/mol) 211.08
MDL Number MFCD00039294
InChI Key OHABWQNEJUUFAV-UHFFFAOYSA-N
Synonym 3,3,3-trifluoropropyl methyldichlorosilane, 3,3,3-trifluoropropyl dichloromethylsilane, dichloro methyl 3,3,3-trifluoropropyl silane, unii-38xgw8m24k, silane, dichloromethyl 3,3,3-trifluoropropyl, dichloromethyl-3,3,3-trifluoropropylsilane, dichloromethyl 3,3,3-trifluoropropyl silane, dichloro-methyl-3,3,3-trifluoropropyl silane, 4-04-00-04169 beilstein handbook reference, ksc491g7h
PubChem CID 12666
IUPAC Name dichloro-methyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](CCC(F)(F)F)(Cl)Cl
Specifications
Boiling Point 121°C to 122°C
Quantity 5 g
UN Number UN2985
Beilstein 1744964
Flash Point 15°C (59°F)
Formula Weight 211.09
Percent Purity 97%
Refractive Index 1.385
Sensitivity Moisture sensitive
Density 1.261
Chemical Name or Material (3,3,3-Trifluoropropyl)methyldichlorosilane