Learn More
3,3'-Thiodipropionic acid, 99%, Thermo Scientific Chemicals
154.26 DKK - 1192.22 DKK
Chemical Identifiers
CAS 111-17-1
Molecular Formula C6H10O4S
Molecular Weight (g/mol) 178.2
MDL Number MFCD00002781
InChI Key ODJQKYXPKWQWNK-UHFFFAOYSA-N
Synonym 3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
PubChem CID 8096
IUPAC Name 3-(2-carboxyethylsulfanyl)propanoic acid
SMILES C(CSCCC(=O)O)C(=O)O
View More Specs
Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10205000
SDS View Documents
Thermo Scientific Acros
138750050
5 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10183710
SDS View Documents
Thermo Scientific Acros
138750250
25 g Plastic bottle
162.71 DKK
25g
10295040
SDS View Documents
Thermo Scientific Acros
138755000
500 g Plastic bottle
335.92 DKK
500g
10391334
SDS View Documents
Thermo Scientific Acros
138750025
2.5 kg Plastic bottle
1192.22 DKK
2.50kg
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Identifiers
111-17-1
178.2
ODJQKYXPKWQWNK-UHFFFAOYSA-N
8096
C(CSCCC(=O)O)C(=O)O
C6H10O4S
MFCD00002781
3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
3-(2-carboxyethylsulfanyl)propanoic acid
Specifications
111-17-1
100.0
White
Authentic
C6H10O4S
MFCD00002781
15, 9485
Solubility in water: 40g/L (25°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol and acetone
C(CSCCC(=O)O)C(=O)O
178.2
178.2
Fine Crystalline Powder
98.5
130.0°C to 133.0°C
128°C
99%
S(CH2CH2CO2H)2
03, IV, 735
3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
ODJQKYXPKWQWNK-UHFFFAOYSA-N
3-(2-carboxyethylsulfanyl)propanoic acid
8096
99%
3,3′-Thiodipropionic acid, 99%
Videos
Safety and Handling
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-841-3
missing translation for 'rtecsNumber' : UF7990000
missing translation for 'tsca' : TSCA
Product Identifier
  • 3,3``-Thiodipropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.