Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve, ISO Certified, Pore Sizes: 10 mm and Larger
Shop All Retsch Products
Click to view available options
Pore Size:
4 mm
10 mm
11.20 mm
12.50 mm
13.20 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.40 mm
25 mm
26.50 mm
28 mm
31.50 mm
35.50 mm
37.50 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
Specifications
Specifications
Quantity | 1Each |
Pore Size | 106 mm |
Type | Test Sieve |
Certifications/Compliance | ISO 3310/1 |
Material | Stainless Steel |
For Use With (Equipment) | Shaker AS 300 |
Includes | Certificate indicating Compliance |
Height (Metric) | 40mm |
For Use With (Application) | For separation, fractioning, particle size determination |
Diameter (Metric) Outer | 305 mm |