Learn More
3M™ PELTOR™ Hygiene Kit for X Series Ear Muffs
3M™ PELTOR™ Hygiene Kit includes replaceable ear cushions and foam liners for X Series Ear Muffs
98.15 DKK - 136.00 DKK
Specifications
Cushion Color | Black |
---|---|
For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
Product Type | Hygiene Kit |
Dielectric | Yes |
Product Code | Brand | Model | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | For Use With (Equipment) | Price | Quantity & Availability | ||||
13439928
|
3M™
7000104045 |
HYX1 | PELTOR™ X1 Ear Muffs |
106.00 DKK
Each |
Estimated Shipment: 07-11-2025
Log in to see stock. |
||||
13449928
|
3M™
7000104046 |
HYX2 | PELTOR™ X2 Ear Muffs |
98.15 DKK
Each |
Estimated Shipment: 07-11-2025
Log in to see stock. |
||||
13459928
|
3M™
7000104047 |
HYX3 | PELTOR™ X3 Ear Muffs |
110.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||
13469928
|
3M™
7000104048 |
HYX4 | PELTOR™ X4 Ear Muffs |
129.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||
13479928
|
3M™
7000104049 |
HYX5 | PELTOR™ X5 Ear Muffs |
136.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||
Description
- Hygiene kits consist of one pair of replacement cushions and foam inserts for 3M™ PELTOR™ X Series ear muffs (X1, X2, X3, X4, X5)
- 3M recommends replacing ear muff cushions and foam inserts every six months or whenever they become damaged.
- Kits are packaged 10 Each/Cs.
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Black | |
Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding | |
Hygiene Kit | |
Yes |