Learn More
4-Benzyloxybenzeneboronic acid, 97%, Thermo Scientific™
Product Code. 11485777
Click to view available options
Quantity:
0.25 g
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as electronic intermediate. It is also utilized in HPLC and NMR spectroscopy.
Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 146631-00-7
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.054
MDL Number MFCD01075705
InChI Key DMJHEIDWSIAXCS-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylboronic acid, 4-benzyloxybenzeneboronic acid, 4-benzyloxy phenyl boronic acid, 4-benzyloxy phenylboronic acid, 4-benzyloxyphenyl boronic acid, 4-benzyl-oxy phenyl boranediol, boronic acid, 4-phenylmethoxy phenyl, 4-phenylmethoxyphenyl boronic acid, pubchem9561
PubChem CID 2734314
IUPAC Name (4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O
Specifications
Melting Point 193°C to 198°C
Quantity 1 g
Formula Weight 228.06
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybenzeneboronic acid