missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorocinnamic acid, 98%, Thermo Scientific™

Product Code. p-7023056
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-71-2
Molecular Formula C9H6BrFO2
Molecular Weight (g/mol) 245.047
MDL Number MFCD00143427
InChI Key ROTBALNVXDGIQY-DAFODLJHSA-N
Synonym 5-bromo-2-fluorocinnamic acid, 2e-3-5-bromo-2-fluorophenyl prop-2-enoic acid, 3-5-bromo-2-fluorophenyl prop-2-enoic acid, 3-5-bromo-2-fluoro-phenyl-acrylic acid, e-3-5-bromo-2-fluorophenyl acrylic acid, 3-5-bromo-2-fluorophenyl-2-propenoic acid, pubchem3230, rarechem bk hw 0189, timtec-bb sbb006748, attercop-chm at116397
PubChem CID 688316
IUPAC Name (E)-3-(5-bromo-2-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1Br)C=CC(=O)O)F

Specifications

Melting Point 193°C to 196°C
Quantity 5 g
Formula Weight 245.05
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluorocinnamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.