Learn More
Greiner Bio-One 5 mL Sterile Serological Pipets
GREENER_CHOICE
Drip-free pipetting design with high optical clarity and maximum accuracy.
Brand:  Greiner Bio-One 606160
 View more versions of this product
Product Code. 13002013
  • 57.14 € / Pack of 200
Only null left
Estimated Shipment: 23-07-2025
Description
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shape: Standard
Sterile
1/10
5 mL
Polystyrene
Yes
Individual Plastic Wrap
Serological Pipet