Learn More
Sigma Laborzentrifugen™ 6-16HS Centrifuge
Suitable for low-speed operation (e.g. for cell centrifugation). Sigma Laborzentrifugen 6-16HS Centrifuge is compatible with various swing-out and fixed-angle rotors.
Brand: Sigma Laborzentrifugen™ 10393
This item is not returnable.
View return policy
Description
- Universal ambient benchtop centrifuge
- Heatable version
- Used specially for ASTM crude oil centrifugation
- Maximum rotor temperature +70°C
- Speed range up to 13,500 rpm
- User-friendly Spincontrol S controller
- Backlit Start, Stop, and Open Lid buttons
- Capacity: 4 x 800mL
- Zero-maintenance induction motor
- Two motorised lid locks
- Magnetic rotor identification
- Stainless-steel centrifuge bowl
- Motorised lid lock
- Easy lid opening with gas spring assistance
- Produced in compliance with latest national and international standards (EN 61010-2-020)
Specifications
4 x 800 mL | |
6-16HS | |
220/240 V 50/60 Hz |
13,500 rpm | |
483 x 581 x 711 mm |