Learn More
SHOWA™ 7585 Single-Use Nitrile Gloves, Powder-Free
Heavy duty, extra strong, highly penetration safe, long cuff single use chemical protective glove!
209.00 DKK
Specifications
Certifications/Compliance (Europe) | EU Regulation 425/2016, EN 420, EN 455, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011 |
---|---|
AQL | 0.65 |
Color | Cobalt Blue |
Disposable | Single-use |
Hand | Ambidextrous |
Description
- Thicker glove provides more resistance to chemicals
- Longer cuff for more protection
- Avoids latex allergies risks type I and type IV
- Second skin feel, softer texture and easy donning
- Low-modulus formulation to improve fit and reduce hand fatigue
- Textured finish on fingertips to enhance grip
- Without plasticisers, powder-free and silicone-free
Specifications
EU Regulation 425/2016, EN 420, EN 455, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011 | |
Cobalt Blue | |
Ambidextrous | |
Nitrile | |
Non-sterile | |
Textured | |
30 cm |
0.65 | |
Single-use | |
Yes | |
Powder Free | |
0.20 mm | |
dispenser box |