Learn More
Testo™ Accurate Immersion/Penetration Probe
Brand:  Testo™ 0628 0006
Product Code. 10593492
  • 1005.00 DKK / Each
Only null left
Estimated Shipment: 01-08-2025
Specifications
Immersion/Penetration Probe
±0.2°C (–25 to 75.0°C), ±0.4°C (–35 to –25.1°C), ±0.4°C (75 to 80°C)
5 sec.
-35°C to +80°C