Learn More
Dostmann Electronic Adapter Plug
Rich, robust, reliable adapter for connecting type K probes with miniature connector on P600/​T900-series
Brand:  Dostmann Electronic 5600-0048
Product Code. 10068581
  • 301.00 DKK / Each
Estimated Shipment: 04-06-2025
Explore more special offers
Description
  • Adapter DIN connector to type K miniature connector
  • Adapter for connecting type K probes with miniature connector on P600/​T900-series
  • Dimensions:100 mm cable length
  • Weight: 25 g
Specifications
Specifications
Type K Thermocouple
Adapter Plug
Documents
One moment while we fetch your results.
Product Title
Dostmann Electronic Adapter Plug > Probe Type: K