missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose, MP Biomedicals™
Agarose Genetic Technology Grade, White Powder
3610.00 DKK
Chemical Identifiers
CAS | 9012-36-6 |
---|---|
Molecular Formula | C24H38O19 |
Molecular Weight (g/mol) | 630.55 |
MDL Number | MFCD00081294 |
InChI Key | MJQHZNBUODTQTK-WKGBVCLCSA-N |
Synonym | 3,6-Anhydro-a-L-galacto-b-D-galactan, Sepharose |
Description
- Agarose is used to separate nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations.
- Agarose gels tend to be more solid, but less elastic, than cross-linked acrylamide gels.
- Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
- Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
Chemical Identifiers
9012-36-6 | |
630.55 | |
MJQHZNBUODTQTK-WKGBVCLCSA-N |
C24H38O19 | |
MFCD00081294 | |
3,6-Anhydro-a-L-galacto-b-D-galactan, Sepharose |
Specifications
Powder | |
87.3°C to 89.5°C | |
5.5 to 7.5 (1.5% aq. soln.) | |
35 - 37°C | |
C24H38O19 | |
3,6-Anhydro-a-L-galacto-b-D-galactan, Sepharose | |
MJQHZNBUODTQTK-WKGBVCLCSA-N | |
Genetic Technology |
9012-36-6 | |
White | |
0.11 - 0.17 | |
> 2000gm/cm2(1.5%gel) | |
MFCD00081294 | |
1g in 100 mL, in water. | |
630.55 |
Safety and Handling
missing translation for 'storageNote' : Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title