Learn More
Al-23 Tube for Heating Coils, O.D.: 60mm, I.D.: 47mm, Pitch: 5.0, Thermo Scientific™
4883.00 DKK - 16623.00 DKK
Specifications
Chemical Name or Material Al-23 Tube for Heating Coils
View More Specs
Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387518
View Documents
Thermo Scientific Alfa Aesar
033210.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11367518
View Documents
Thermo Scientific Alfa Aesar
033210.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11343168
View Documents
Thermo Scientific Alfa Aesar
033210.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11377518
View Documents
Thermo Scientific Alfa Aesar
033210.ED
500 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Tube for Heating Coils
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures