Learn More
Allyl vinyl ether, 95% stab. with 0.1% potassium hydroxide, Thermo Scientific Chemicals
CAS: 3917-15-5 | C5H8O | 84.12 g/mol
780.00 DKK - 3055.00 DKK
Chemical Identifiers
CAS | 3917-15-5 |
---|---|
Molecular Formula | C5H8O |
Molecular Weight (g/mol) | 84.12 |
MDL Number | MFCD00039827 |
InChI Key | ZXABMDQSAABDMG-UHFFFAOYSA-N |
Synonym | allyl vinyl ether, ether, allyl vinyl, vinyl allyl ether, 1-propene, 3-ethenyloxy, allyl ethenyl ether, unii-fj2j0n55cs, fj2j0n55cs, allylvinylether, acmc-20alul, 3-ethenoxy-1-propene |
PubChem CID | 221523 |
IUPAC Name | 3-ethenoxyprop-1-ene |
SMILES | C=CCOC=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348036
|
Thermo Scientific Alfa Aesar
L13320.09 |
10 g |
780.00 DKK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358036
|
Thermo Scientific Alfa Aesar
L13320.18 |
50 g |
3055.00 DKK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Allyl vinyl ether is used as a intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllyl vinyl ether is used as a intermediate in chemical research.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, ultraviolet radiation and free radical initiators.
Chemical Identifiers
3917-15-5 | |
84.12 | |
ZXABMDQSAABDMG-UHFFFAOYSA-N | |
221523 | |
C=CCOC=C |
C5H8O | |
MFCD00039827 | |
allyl vinyl ether, ether, allyl vinyl, vinyl allyl ether, 1-propene, 3-ethenyloxy, allyl ethenyl ether, unii-fj2j0n55cs, fj2j0n55cs, allylvinylether, acmc-20alul, 3-ethenoxy-1-propene | |
3-ethenoxyprop-1-ene |
Specifications
3917-15-5 | |
65°C to 68°C | |
Characteristic | |
CH2=CHCH2OCH=CH2 | |
MFCD00039827 | |
1697665 | |
Slightly soluble in water. | |
C=CCOC=C | |
84.12 | |
84.12 | |
Allyl vinyl ether, stabilized with 0.1% potassium hydroxide |
0.781 | |
<−5°C (23°F) | |
C5H8O | |
1.412 | |
UN3271 | |
allyl vinyl ether, ether, allyl vinyl, vinyl allyl ether, 1-propene, 3-ethenyloxy, allyl ethenyl ether, unii-fj2j0n55cs, fj2j0n55cs, allylvinylether, acmc-20alul, 3-ethenoxy-1-propene | |
ZXABMDQSAABDMG-UHFFFAOYSA-N | |
3-ethenoxyprop-1-ene | |
221523 | |
95% |
Safety and Handling
GHS H Statement
H225-H302-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
missing translation for 'einecsNumber' : 223-482-6
missing translation for 'rtecsNumber' : KM9450000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.