Learn More
Haldenwanger™ Aluminum Ceramic Conical Tall Form Crucibles
164.00 DKK - 1830.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10748361
|
Haldenwanger™
L ALSINTTIEGEL*C/00 |
10 mL |
164.00 DKK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
10232121
|
Haldenwanger™
L ALSINTTIEGEL*C/1 |
20 mL |
393.00 DKK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
10303821
|
Haldenwanger™
L ALSINTTIEGEL*C/2 |
30 mL |
481.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
|||||
10579671
|
Haldenwanger™
L ALSINTTIEGEL*C/3 |
45 mL | N/A | ||||||
10729131
|
Haldenwanger™
L ALSINTTIEGEL*C/6 |
250 mL |
1830.00 DKK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
Specifications
Aluminum Ceramic | |
Crucible | |
Conical |