Learn More
Tenak™ Aluminum Eco AluRack for Chest Freezers
Providing a whole new way of making storage racks. Tenak™ Aluminum Eco AluRacks for Chest Freezers fit into any freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11566
549.80 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Made of seawater resistant aluminum
- Available in a range of box heights
- Racks can be used with all freezer brands
- Sold without cryoboxes
Specifications
Eco AluRack | |
Aluminum | |
For chest freezers | |
1 Pack | |
140 mm | |
32 mm |
Boxes (96) | |
Yes | |
1 Pack | |
140 x 140 x 635mm | |
635 mm | |
140 mm |