Learn More
Invitrogen™ Ambion™ 0.5-10kb RNA Ladder
Suitable for sizing single-stranded RNA ranging from 0.5kb to 10kb in glyoxal or formaldehyde agarose gels
Brand:  Invitrogen™ 15623200
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11528766
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: 0.5-10kb RNA RNA Ladder is supplied at 1μg/μL in 10mM HEPES (pH 7.2), 2mM EDTA
Description
Description
  • Consists of nine bands in even increments (0.5, 1, 1.5, 2, 3, 4, 6, 8, and 10kb) to simplify size estimation
  • Compared to other bands, 1.5kb band is more intense, allowing easy orientation and identification of RNA bands on gel
  • RNA molecules in ladder are polyA-tailed (40 nt) for convenient use as control templates for cDNA synthesis
  • Compatible with formaldehyde-containing, glyoxal-containing and polyacrylamide gels
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
75 μL