Learn More
Thermo Scientific™ ART™ X800 Pure MBP 1000G Genomic Pipette Tip
ART tips provide a quick and convenient guide to monitor the desired volume of liquid
Brand: Thermo Scientific™ 3591
397.60 DKK valid until 2025-12-31
Use promo code "24081" to get your promotional price.
This item is not returnable.
View return policy
Description
- Material: Polypropylene
- Volume: 100 to 1000μL
- Sterile: Yes
- Color: White
- Tip Style: Wide Orifice
- Model: Pure 1000G
- Packaging Type: Rack
Specifications
100 to 1000 μL | |
White | |
Rack | |
ART |
Sterile | |
Pure 1000G | |
Genomic/Wide Orifice | |
Polypropylene |