Learn More
Axygen™ AxyMats™ Sealing Mat for 750uL 96 Round Well Plate and ELISA Plate
Shop All Axygen Scientific Products
Click to view available options
Material:
Silicone
Unit Size:
Pack of 50
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Specifications
Color | Natural |
Material | Silicone |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
No. of Wells | 96 |
No. per Case | 50/Cs. |
No. per Pack | 10/Pk. |
Product Type | Sealing Mat |
Well Shape | Round |
Plate Compatibility | 96-well Round Well Plate and ELISA Plate, 750 μL |