Learn More
Azlon™ Multi-lingual Round Integral Wash Bottle
Azlon™ Multi-lingual Round Integral Wash Bottle is molded with an integral spout to reduce contamination caused by the inner tube touching the benchtop during re-filling.
277.00 DKK - 317.00 DKK
Specifications
Capacity (Metric) | 500 mL |
---|---|
Includes | Integral spout |
Closure Included | Yes |
Mouth | Wide |
Product Type | Wash Bottle |
Product Code | Brand | Chemical Label | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Chemical Label | Price | Quantity & Availability | |||||
15888467
|
Azlon
WGW801P |
Distilled Water |
277.00 DKK
Pack of 5 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
15898467
|
Azlon
WGW802P |
Acetone |
317.00 DKK
Pack of 5 |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
15808477
|
Azlon
WGW803P |
Methanol |
311.00 DKK
Pack of 5 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
15818477
|
Azlon
WGW804P |
Isopropanol |
311.00 DKK
Pack of 5 |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
15828477
|
Azlon
WGW805P |
Ethanol |
317.00 DKK
Pack of 5 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
Description
- Multilingual
- Wide neck wash bottle
- Indelibly printed in English, French, German and Spanish
- Choose from five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
- Fitted with color coded one-piece PP closure that maintains it's shape in use
- Designed with integral spout to prevent contamination
Specifications
500 mL | |
Yes | |
Wash Bottle |
Integral spout | |
Wide | |
Polypropylene |