Learn More
Azlon™ Polyethylene Stoppers
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. Azlon™ Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
Brand:  Azlon™ TRF593
 View more versions of this product
Product Code. 11317804
  • 158.00 DKK / Pack of 10
Explore more special offers
Description
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. AzlonTM Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
  • Manufactured from polyethylene
  • Available in white and red
  • Suitable for use with both Azlon and glass volumetric ware
  • Enlarged head provides a good grip and protects glassware
  • Ribbed cone ensures a secure leak free seal
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glass or Plastic volumetric flasks and measuring cylinders etc
White, Red
Tapered Stopper
14/23
Polyethylene
Ribbed Cone