Learn More
BD Difco™ Dehydrated Culture Media: CzaPEK Solution Agar
For cultivation of organisms capable of utilizing inorganic nitrogen. BD Difco™ Dehydrated Culture Media: CzaPEK Solution Agar is the perfect choice.
Brand: BD 233910
This item is not returnable.
View return policy
Description
- Microbiology media
- Dehydrated
- Solution Agar
- 500g
Specifications
SMWW | |
Dehydrated | |
Agar | |
Cultivation of organisms capable of utilizing inorganic Nitrogen | |
500 g |
2°C to 8°C | |
CzaPEK Solution Agar | |
Saccharose 30g/L, Sodium Nitrate 2g/L, Dipotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Potassium Chloride 0.5g/L, Ferrous Sulfate 0.01g/L, Agar 15g/L | |
Bottle |