Learn More
Sampling Systems™ Beaker with Spout
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
100 mL
200 mL
500 mL
1 L
2 L
3 L
5 L
Description
- Made from high grade 316L stainless steel
- Wide range of sizes available
Specifications
Specifications
Capacity (Metric) | 5 L |
Material | Stainless Steel (316L) |
Graduated | Yes |
Certifications/Compliance | FDA |
Style | Tall Form |