Learn More
Bel-Art™ Cover Glass Forceps
Tips shaped to easily lift cover glass. Bel-Art™ Cover Glass Forceps are fully coated with brown Teflon™ FEP and comply with FDA regulations concerning direct food contact.
Brand: Bel-Art™ H37943-0000
525.68 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Brown PTFE FEP coating guards against scratches
- Wide, bent tip style is perfect for lifting cover glasses
Specifications
Straight | |
PTFE | |
Forceps | |
Lifting cover glasses | |
Slide Holding |
4.5 in. | |
114 mm | |
Bbrown PTFE-FEP coating | |
FEP Coated Cover Glass Forceps |