Learn More
Bel-Art™ Nylon Screw Compressor Clamp
Tubing clamp with angled jaws prevents tube from slipping out
Brand:  Bel-Art™ F18225-0000
Product Code. 10254491
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Tubing clamp with angled jaws prevents tube from slipping out
  • Open-jaw nylon tubing clamp fits any tubing up to 1/4 in. O.D. (6.4mm)
  • Thumbscrew clamp applies even force
Specifications
Specifications
12 Pk.
Nylon
2.31 in.
5.867 cm
Brown
6.4 mm
Screw Compressor Clamp
0.25 in. O.D. Tube
0 to 0.25 in.
0 to 0.63 cm
Angled