Learn More
Bel-Art™ Slide Boxes
Shop All SP Scienceware Products
Click to view available options
Slide Capacity:
100
Unit Size:
Each
Description
- Heavy-walled boxes are rugged and will not bend, split or chip
- Boxes are unaffected by humidity and moisture and are thoroughly insect-proof
- Easily identify slides with numbered index included on base and cover
Specifications
Specifications
Type | Slide Box |
Slide Capacity | 100 |
Device Holds | 100 |
For Use With (Equipment) | Microscope Slide |
Includes | Numbered index on base |