Learn More
Bel-Art™ Spinbar™ Magnetic Sink Strainer
Resistant to chemicals sink strainer has four strong magnets in each corner to attract and hold Spinbar magnetic stirring bars.
Brand: Bel-Art™ F37787-0001
298.26 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- Long slits in the sides and skirt allow fluids to drain easily and rapidly
- A wide skirt enables strainer to be used in a variety of sink drains; flat edge on one side allows it to be placed close to the sink wall
- Four legs on the underside anchor the strainer firmly in sink drain and prevent it from being dislodged by flowing liquid
- Color choices improve visibility of stirring bars
Specifications
Green | |
Prevents stir bar from washing down drain | |
4 strong magnets in each corner, long slits in the sides and wide skirts | |
127 mm | |
1 | |
Magnetic Stir Bar Retriever |
5 x 1.44 in. | |
Stirring bars | |
127 mm | |
PTFE, Alnico Magnet | |
Disc |