Learn More
Bel-Art™ Sterileware™ Economy Sample Spoons
Molded high-impact polystyrene for durability and stiffness. Ideal for retrieving small, specific volume samples of powders or liquids.
Brand: Bel-Art™ F36740-0002
944.30 DKK valid until 2025-08-31
Use promo code "24124" to get your promotional price.
This item is not returnable.
View return policy
Description
- Molded of durable, white high-impact polystyrene for durability and stiffness when sampling packed powders
- Flat-bottom shape permits use as a weighing boat
- For one-time use
Order Info
100 individually sterile wrapped spoons or 12 nonsterile spoons per pack
Specifications
Spoon | |
Not Autoclavable | |
Flat | |
Single-use | |
White | |
Designed for retrieving small, specific volume samples of powders or liquids and also provides durability and stiffness when sampling packed powders |
Polystyrene | |
Certificate of processing for sterilization | |
2.5 mL | |
Sterile | |
FDA | |
100/Pk. |