Learn More
Lonza™ BioWhittaker™ Dulbecco's Phosphate Buffered Saline (1X) w/o Calcium and Magnesium
Used to provide buffer system to maintain cell culture media in the physiological pH range of 7 to 7.6.
143.00 DKK
Specifications
Product Type | Dulbecco's Phosphate Buffered Saline |
---|---|
Without Additives | No Calcium, No Magnesium |
pH | 7 to 7.6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10110035
|
Lonza™
BE17-512F |
500 mL | N/A | N/A | N/A | ||||
Description
- Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
- Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
Specifications
Dulbecco's Phosphate Buffered Saline | |
No Calcium, No Magnesium | |
7 to 7.6 |