Learn More
CellPath BiTran™ High Visibility Barrier Film Specimen Bag
Leak proof bags for the safe transportation of specimens
Brand: CellPath WAA-0500-12A
This item is not returnable.
View return policy
Description
- For transporting wet tissue from one place to another
- Available in six sizes
- Barrier film impermeable to formalin vapour
- Double zip lock seal provides leak proof closure
Specifications
Zip Seal | |
High-visibility Barrier Film | |
230 x 300mm |
For transporting wet tissue, short to long-term storage of tissue | |
250 Pack | |
Specimen Bag |