Learn More
Helios Italquartz Boats with Ring Handle, Transparent
Click to view available options
Dimensions (L x W):
120 mm x 15 mm
143 mm x 17 mm
180 mm x 15 mm
76 mm x 10 mm
76 mm x 11 mm
Unit Size:
Each
Description
- Transparent
- Include ring handle
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Specifications
Bottom Shape | Flat |
Color | Transparent |
Material | Quartz |
Shape | Boat |
Type | Weighing Boat with Ring Handle |
Dimensions (L x W) | 76 mm x 10 mm |
Length (Metric) | 76/16 mm |
Width (Metric) | 10 mm |
Height (Metric) | 10 mm |