Learn More
Retsch™ Bottom Sieves SK 300 with Trapezoid Holes
Retsch™ Bottom Sieves SK 300 with Trapezoid Holes are constructed of stainless steel for durability and suitable for use with Cross Beater Mill SK 300.
1810.00 DKK
Specifications
Description | Trapezoid holes |
---|---|
Type | Sieve |
Material | Stainless Steel |
For Use With (Equipment) | Cross Beater Mill SK 300 |
Description
- Stainless steel
- Trapezoid holes
- For use with cross beater
Specifications
Trapezoid holes | |
Sieve | |
Stainless Steel | |
Cross Beater Mill SK 300 |