Learn More
BRAND™ QuikSip™ Bottle Top Aspirator Accessory, Manifold
Autoclavable and used withQuikSip™ Bottle Top Aspirator
Brand: BRAND™ 704526
85.42 DKK valid until 2025-08-31
Use promo code "24124" to get your promotional price.
This item is not returnable.
View return policy
Description
- 8-channel
- Polypropylene
- Autoclavable (121°C)
Specifications
8 Channel Manifold | |
Polypropylene | |
QuikSip bottletop aspirator |
Autoclavable | |
Removal of fluids from multiwell plates |