Learn More
Buffer Solution pH 4.00, Crison Instruments™
Click to view available options
Quantity:
250 mL
Unit Size:
Each
Specifications
Specifications
Melting Point | 0°C |
Color | Undesignated |
pH | 4.00 |
Physical Form | Liquid |
Quantity | 250 mL |
Formulation | Potassium Hydrogen Phthalate, also contains Germicide |
Odor | Odorless |
Traceability to NIST | Yes |
Solubility Information | Complete soluble in water |