Learn More
Merck Millipore BugBuster™ 10X Protein Extraction Reagent
Reagent for protein sample preparation
Brand: Merck Millipore 70921-4
4170.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Concentrated formulation of proprietary detergents without addition of salts or buffer components
- Provides flexible alternative to ready-to-use standard 1X BugBuster, allowing user-defined dilution and addition of buffer components
- Consists all of the bioprocessing benefits of standard BugBuster plus the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein