Learn More
Pal International™ Bouffant Caps
Manufactured from soft, non-woven material. Pal International™ Bouffant Caps are larger than a mob cap and provide completed hair covering for those with longer hair.
651.00 DKK - 715.00 DKK
Specifications
Product Type | Bouffant |
---|---|
Size | One Size |
Material | Polypropylene |
Disposable | Single-use |
Latex Free | Yes |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12313996
|
Pal International™
D84110 |
White |
651.00 DKK
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||||
12323996
|
Pal International™
D84230 |
Blue |
715.00 DKK
Pack of 1000 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
Description
- Soft, light non-woven polypropylene with an elasticated edge
- Complete hair coverage
- Ultra low-linting, making them ideally suited to working environments where contamination by loose fibers may be a risk
- Food safe and Latex-free for reduced irritation
- Larger than a mob cap and designed for longer hair
- One size fits all
Specifications
Bouffant | |
Polypropylene | |
Yes |
One Size | |
Single-use | |
1000/Pk. |