missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf45 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | C16orf45 |
---|---|
Concentration | 0.49 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16864565
|
Proteintech
26037-1-AP-20UL |
20 μL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16854565
|
Proteintech
26037-1-AP-150UL |
150 μL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
C16orf45 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q96MC5 | |
360459, 89927 | |
C16orf45 Fusion Protein Ag22584 | |
Primary | |
-20°C | |
C16orf45 |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C16orf45 | |
C16orf45, RGD1305733 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title