Learn More
Uvex™ C300 Dry Cut Protection Gloves
Uvex C300 Dry gloves are designed to provide level 3/C cut protection and feature HPV dots on the palm for enhanced grip capabilities.
91.30 DKK
Specifications
Coating Material | HPV Dots |
---|---|
Color | Gray, Green |
Material | Bamboo Viscose, HPPE, Polyamide, Glass |
EN388 Standard | XX4XC |
Glove Cuff Style | Knit |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15357102
|
Uvex™
6054908 |
8 | N/A | ||||||
Description
- Gloves provide level 3/C blade cut resistance, and very high tear resistance
- High-performance vinyl (HPV) dots on the palm enhance grip capabilities in dry environments
- Uvex Climazone technology minimizes excessive warmth and moisture
- Highly flexible with very good tactile sensation
- Silicone-free according to the imprint test
- 3D Ergo technology ensures a secure fit
Specifications
HPV Dots | |
Bamboo Viscose, HPPE, Polyamide, Glass | |
Knit |
Gray, Green | |
XX4XC | |
C300 Dry |