Learn More
Calcium Sulfate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Product Code. 10650331 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
1 kg
Chemical Identifiers
CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number 149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O
Specifications
Color White
pH 7
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 172.17g/mol
Packaging Plastic powder jar