Learn More
Erlab™ Captair™ Mobicap™ Rolling Carts
Specifications
Height (Metric) | 881 mm |
---|---|
Depth (Metric) | 600 mm |
Product Type | Mobicap™ Rolling Cart |
Material | Metallic Alloy |
Specifications
881 mm | |
600 mm | |
Mobicap™ Rolling Cart | |
Metallic Alloy |