Learn More
Carl Zeiss™ 100W Halogen Rectangular Filament Bulb
Providing excellent illumination for halogen lamps in the laboratory.
Brand:  Carl Zeiss™ 380059-1660-000
Product Code. 10707070
  • 21.70 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Description
  • 12 volts, 100 watts
  • For use in halogen lamps
Specifications
Specifications
Microscope Bulb
Rectangular
100 w
Standard WL Universal, photomikroskop III, IM 35, ICM 405, Axiom at UEM, ACM stand U in lamp housing 100/250, Axioplan (1986) lamp housing 100 No. 467259-9901/447215
12 V