Learn More
CCDC6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13717-1-AP-150UL
 View more versions of this product
Product Code. 16852074
  • 3058.99 DKK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.
Specifications
Specifications
CCDC6
Polyclonal
Unconjugated
CCDC6
CCDC6, D10S170, H4, Protein H4, PTC, TPC, TST1
Rabbit
Antigen Affinity Chromatography
RUO
8030
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16204
CCDC6
CCDC6 Fusion Protein Ag4658
150 μL
Primary
Human
Antibody
IgG