missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13717-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.Specifications
CCDC6 | |
Polyclonal | |
Unconjugated | |
CCDC6 | |
CCDC6, D10S170, H4, Protein H4, PTC, TPC, TST1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8030 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16204 | |
CCDC6 | |
CCDC6 Fusion Protein Ag4658 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction